U3879
Uracil-2-14C
aqueous solution (containing 2% ethanol)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H4N2O2
CAS Number:
Molecular Weight:
112.09
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
aqueous solution (containing 2% ethanol)
extent of labeling
45-60 mCi per mmol
mass shift
M+2
storage temp.
2-8°C
SMILES string
O=C1N[14C](=O)NC=C1
Packaging
serum bottle
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service