Key Documents
TMO4113-0250
Nalgene® sterile single-use Erlenmeyer flask with solid closure
PETG, bottle volume 250 mL, baffled bottom, case of 12 ea
Synonym(s):
Nalgene Erlenmeyer flask, single use plastic flask
Select a Size
1 EACH
€208.00
€208.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
€208.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.13
€208.00
Please contact Customer Service for Availability
material
PETG
sterility
sterile
packaging
case of 12 ea
manufacturer/tradename
Nalgene 4113-0250
bottle volume
250 mL
General description
Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Other Notes
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)