Key Documents
TMO312087-0008
Nalgene® narrow-mouth bottle
natural polypropylene copolymer, size 250 mL, case of 250 ea
Synonym(s):
Nalgene bottle, plastic bottle
Select a Size
250 EACH
€530.00
€530.00
Please contact Customer Service for Availability
Select a Size
Change View
250 EACH
€530.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€530.00
Please contact Customer Service for Availability
material
natural polypropylene copolymer
packaging
case of 250 ea
manufacturer/tradename
Nalgene 312087-0008
size
250 mL
General description
Store and ship light-sensitive reagents with Thermo Scientific Nalgene Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)