Skip to Content
Merck
All Photos(1)

Key Documents

T161

Supelco

Testosterone isocaproate

Synonym(s):

4-Androsten-17β-ol-3-one 17-(4-methylpentanoate), Testosterone 4-methylvalerate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H38O3
CAS Number:
Molecular Weight:
386.57
EC Number:
MDL number:
UNSPSC Code:
41116107

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])OC(=O)CCC(C)C

InChI

1S/C25H38O3/c1-16(2)5-10-23(27)28-22-9-8-20-19-7-6-17-15-18(26)11-13-24(17,3)21(19)12-14-25(20,22)4/h15-16,19-22H,5-14H2,1-4H3/t19-,20-,21-,22-,24-,25-/m0/s1

InChI key

PPYHLSBUTAPNGT-BKWLFHPQSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Büyükgebiz
The Turkish journal of pediatrics, 35(4), 271-275 (1993-10-01)
Thirteen boys with constitutional delayed puberty (CDP) were treated with a combination of short and long-acting testosterone esters (testosterone propionate, testosterone phenylpropionate, testosterone isocaproate). Mean age at the onset of treatment was 14.9 +/- 0.6 years and bone age delay
A Büyükgebiz
Hormone research, 44 Suppl 3, 32-34 (1995-01-01)
Thirteen boys who had constitutional delayed puberty (CDP) were treated with a combination of short- and long-acting testosterone esters (testosterone propionate, testosterone phenylpropionate, testosterone isocaproate). Mean age at the onset of treatment was 14.9 +/- 0.6 years and bone age

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service