Skip to Content
Merck
All Photos(1)

Key Documents

SRP0101

Sigma-Aldrich

HDAC 1 from Plasmodium falciparum

recombinant, expressed in baculovirus infected insect cells, ≥40% (SDS-PAGE)

Synonym(s):

HD1, Histone deacetylase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

Plasmodium falciparum

recombinant

expressed in baculovirus infected insect cells

Assay

≥40% (SDS-PAGE)

form

aqueous solution

mol wt

55 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Plasmodium falciparum ... HDAC1(813532)

General description

Plasmodium falciparum HDAC1, GenBank Accession No. XM_001352091, full length with C-terminal HIS6-DDDDK (FLAG) tags, MW = 55 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme required to deacetylate 1 pmol of substrate/min at 37°C.

Physical form

Formulated in 50 mM Tris-HCl, pH 8.0, 130 mM NaCl and 10% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service