Skip to Content
Merck
All Photos(2)

Key Documents

SML3914

Sigma-Aldrich

Cabozantinib

≥98% (HPLC)

Synonym(s):

BMS 907351, BMS-907351, BMS907351, N-(4-(6,7-Dimethoxyquinolin-4-yloxy)phenyl)-N’-(4-fluorophenyl)cyclopropane1,1-dicarboxamide, N-(4-{[6,7-bis(Methyloxy)quinolin-4-yl]oxy}phenyl)-N′-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide, XL 184, XL-184, XL184

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€91.50
50 MG
€295.00

€91.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€91.50
50 MG
€295.00

About This Item

Empirical Formula (Hill Notation):
C28H24FN3O5
CAS Number:
Molecular Weight:
501.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€91.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

Fc1ccc(cc1)NC(=O)C5(CC5)C(=O)Nc2ccc(cc2)Oc3c4c(ncc3)cc(c(c4)OC)OC

InChI

1S/C28H24FN3O5/c1-35-24-15-21-22(16-25(24)36-2)30-14-11-23(21)37-20-9-7-19(8-10-20)32-27(34)28(12-13-28)26(33)31-18-5-3-17(29)4-6-18/h3-11,14-16H,12-13H2,1-2H3,(H,31,33)(H,32,34)

InChI key

ONIQOQHATWINJY-UHFFFAOYSA-N

Biochem/physiol Actions

Cabozantinib (XL184; BMS‐907351) is an orally active, potent inhibitor against multiple receptor tyrosine kinases, notably VEGFR2/KDR (IC50 = 35 pM), MET (IC50 = 1.3 nM/wt, 3.8 nM/Y1248H, 11.8 nM/D1246N, 14.6 nM/K1262R), KIT, RET, AXL, TIE2, and FLT3 (IC50 = 4.6, 5.2, 7, 14.3, and 11.3 nM, respectively). Cabozantinib inhibits MET-driven proliferation in cancer cultures (IC50 = 9.9 nM/Hs746T, 19 nM/SNU-5) and simultaneously suppressing metastasis, angiogenesis and tumor growth in vivo (3-100 mg/kg via single or daily p.o. to mice with human MDA-MB-231 xenograft, 1-60 mg/kg via single or daily p.o. to rats with C6 rat glioma).
Orally active, potent receptor tyrosine kinase inhibitor against VEGFR2 (KDR), MET, KIT, RET, AXL, TIE2, and FLT3 with anti-tumor efficacy in vivo.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B - STOT RE 2

Target Organs

Bone marrow,Kidney,Endocrine system,Gastrointestinal tract,Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cabozantinib: A Multitargeted Oral Tyrosine Kinase Inhibitor
Pharmacotherapy, 38(3), 357-369 (2018)
Cabozantinib Enhances Anti-PD1 Activity and Elicits a Neutrophil-Based Immune Response in Hepatocellular Carcinoma
Clinical Cancer Research, 28(11), 2449-2460 (2022)
F Michael Yakes et al.
Molecular cancer therapeutics, 10(12), 2298-2308 (2011-09-20)
The signaling pathway of the receptor tyrosine kinase MET and its ligand hepatocyte growth factor (HGF) is important for cell growth, survival, and motility and is functionally linked to the signaling pathway of VEGF, which is widely recognized as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service