Skip to Content
Merck
All Photos(1)

Key Documents

SML2516

Sigma-Aldrich

Valrubicin

Synonym(s):

Trifluoroacetyladriamycin 14-valerate

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
€219.00

€219.00


Please contact Customer Service for Availability


Select a Size

Change View
20 MG
€219.00

About This Item

Empirical Formula (Hill Notation):
C34H36F3NO13
CAS Number:
Molecular Weight:
723.64
UNSPSC Code:
12352200
NACRES:
NA.77

€219.00


Please contact Customer Service for Availability

form

powder or crystals

storage temp.

2-8°C

SMILES string

FC(F)(F)C(=O)NC1C(C(OC(C1)O[C@H]2C[C@](Cc3c2c(c4c(c3O)C(=O)c5c(c(ccc5)OC)C4=O)O)(O)C(=O)COC(=O)CCCC)C)O

InChI

1S/C34H36F3NO13/c1-4-5-9-21(40)49-13-20(39)33(47)11-16-24(19(12-33)51-22-10-17(27(41)14(2)50-22)38-32(46)34(35,36)37)31(45)26-25(29(16)43)28(42)15-7-6-8-18(48-3)23(15)30(26)44/h6-8,14,17,19,22,27,41,43,45,47H,4-5,9-13H2,1-3H3,(H,38,46)/t14?,17?,19-,22?,27?,33-/m0/s1

InChI key

ZOCKGBMQLCSHFP-ZQUOIQDWSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Valrubicin is a semisynthetic derivative of the antineoplastic anthracycline antibiotic doxorubicin. Valrubicin is converted intracytoplasmically into N-trifluoroacetyladriamycin, which interacts with topoisomerase II, stabilizing the complex between the enzyme and DNA and causing cell cycle arrest.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Muta. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service