Skip to Content
Merck
All Photos(1)

Documents

SML1703

Sigma-Aldrich

2-BFI hydrochloride

≥98% (HPLC)

Synonym(s):

2 BFI, 2-(2-benzofuranyl)-4,5-dihydro-1H-imidazole hydrochloride, 2-(Benzofuran-2-yl)-2-imidazoline hydrochloride, RX 801077

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10N2O · HCl
CAS Number:
Molecular Weight:
222.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[H]Cl.C12=CC=CC=C1OC(C3=NCCN3)=C2

InChI

1S/C11H10N2O.ClH/c1-2-4-9-8(3-1)7-10(14-9)11-12-5-6-13-11;/h1-4,7H,5-6H2,(H,12,13);1H

InChI key

RFNFFVDVGWOSNZ-UHFFFAOYSA-N

Biochem/physiol Actions

2-BFI is a high-affinity Imidzoline I2 receptor ligand (Ki = 9.8 nM) In an in vivo functional assay, 2-BFI induces hypothermia in rats.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service