Skip to Content
Merck
All Photos(1)

Documents

SML1239

Sigma-Aldrich

XL388

≥98% (HPLC)

Synonym(s):

XL-388, [7-(6-Amino-3-pyridinyl)-2,3-dihydro-1,4-benzoxazepin-4(5H)-yl][3-fluoro-2-methyl-4-(methylsulfonyl)phenyl]-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22FN3O4S
CAS Number:
Molecular Weight:
455.50
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

InChI

1S/C23H22FN3O4S/c1-14-18(5-7-20(22(14)24)32(2,29)30)23(28)27-9-10-31-19-6-3-15(11-17(19)13-27)16-4-8-21(25)26-12-16/h3-8,11-12H,9-10,13H2,1-2H3,(H2,25,26)

InChI key

LNFBAYSBVQBKFR-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

XL388 is a potent, selective, orally bioavailable ATP-competitive inhibitor of mammalian target of rapamycin (mTOR) with an IC50 of 9.9 nM for mTOR, 8 nM for mTORC1, 166 nM for mTORC2, and 1000-fold selectivity for mTOR over the closely related PI3K kinases and a panel of 141 protein kinases tested. XL388 blocked mTORC1 phosphorylation of p70S6K (T389) with an IC50 value of 94 nM and blocked mTORC2 phosphorylationof AKT (S473) with an IC50 value of 350 nM. XL388 showed complete tumor growth inhibition in mice bearing MCF-7 xenograft tumors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service