Skip to Content
Merck
All Photos(1)

Key Documents

SML0458

Sigma-Aldrich

Cardionogen-1

≥98% (HPLC)

Synonym(s):

6-Cyclohexyl-3-furan-2-yl-[1,2,4]triazolo[3,4-b][1,3,4]thiadiazole, CDNG1, Cardionogen-1/vuc230

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N4OS
CAS Number:
Molecular Weight:
274.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

C1CCC(CC1)c2nn3c(nnc3s2)-c4ccco4

InChI

1S/C13H14N4OS/c1-2-5-9(6-3-1)12-16-17-11(10-7-4-8-18-10)14-15-13(17)19-12/h4,7-9H,1-3,5-6H2

InChI key

SWTUPSNBTSPBIH-UHFFFAOYSA-N

Biochem/physiol Actions

Cardionogen-1 is cardiomyocyte promoter and an inhibitor of Wnt/β-catenin dependent transcriptional activity with an EC50 of 23 nM in murine stem cells and zebrafish embryos. It induces cardiac cell formation in both murine stem cells and zebrafish embryos, expanding cardiac progenitor cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service