Skip to Content
Merck
All Photos(1)

Documents

SCP0062

Sigma-Aldrich

Calmodulin Dependent Protein Kinase Substrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H80N14O15
Molecular Weight:
1045.19
UNSPSC Code:
12352202
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Pro-Leu-Ser-Arg-Thr-Leu-Ser-Val-Ser-Ser-NH2

Application

Pro-Leu-Ser-Arg-Thr-Leu-Ser-Val-Ser-Ser-NH2 is a calmodulin-dependent protein kinase substrate used to identify, differentiate, quantitate and characterized CaM-dependent protein kinases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R B Pearson et al.
The Journal of biological chemistry, 260(27), 14471-14476 (1985-11-25)
The substrate specificity of the multifunctional calmodulin-dependent protein kinase from skeletal muscle has been studied using a series of synthetic peptide analogs. The enzyme phosphorylated a synthetic peptide corresponding to the NH2-terminal 10 residues of glycogen synthase, Pro-Leu-Ser-Arg-Thr-Leu-Ser-Val-Ser-Ser-NH2, stoichiometrically at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service