SAB5702245
Anti-CD28 Antibody, clone 5G1C8, Rabbit Monoclonal
Synonym(s):
Tp44
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
biological source
rabbit
Quality Level
material
colorless
clone
5G1C8, monoclonal
form
liquid
mol wt
45 kDa
species reactivity
human, rat, rat
concentration
1.00 mg/mL
technique(s)
flow cytometry: 1:50 - 1:200
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CD28(940)
General description
The protein encoded by this gene is essential for T-cell proliferation and survival, cytokine production, and T-helper type-2 development. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Immunogen
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.
Recombinant protein of human CD28.
Application
WB, FC
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service