Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702130

Sigma-Aldrich

Anti-[KD Validated] Casein Kinase 2 alpha (CSNK2A1) Antibody, clone 2P3M7, Rabbit Monoclonal

Synonym(s):

CK2A1,CKII,CSNK2A3,OCNDS,CKII alpha,CSNK2A1,Cka1,Cka2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
2P3M7, monoclonal
application:
WB
species reactivity:
human, rat, rat
technique(s):
western blot: 1:500 - 1:2000
citations:
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

material

colorless

clone

2P3M7, monoclonal

form

liquid

mol wt

42 kDa

species reactivity

human, rat, rat

concentration

0.6mg/ml

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MSGPVPSRARVYTDVNTHRPREYWDYESHVVEWGNQDDYQLVRKLGRGKYSEVFEAINITNNEKVVVKILKPVKKKKIKREIKILENLRGGPNIITLADI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSNK2A1(1457)

General description

Casein kinase II is a serine/threonine protein kinase that phosphorylates acidic proteins such as casein. It is involved in various cellular processes, including cell cycle control, apoptosis, and circadian rhythm. The kinase exists as a tetramer and is composed of an alpha, an alpha-prime, and two beta subunits. The alpha subunits contain the catalytic activity while the beta subunits undergo autophosphorylation. The protein encoded by this gene represents the alpha subunit. While this gene is found on chromosome 20, a related transcribed pseudogene is found on chromosome 11. Three transcript variants encoding two different proteins have been found for this gene. [provided by RefSeq, Jul 2014]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Casein Kinase 2 alpha (CSNK2A1) (P68400).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service