Skip to Content
Merck
All Photos(3)

Key Documents

SAB4702019

Monoclonal Anti-CD82 purified low endotoxin produced in mouse

new

Synonym(s):

4F9, C33, GR15, IA4, KAI1, R2, SAR2, ST6, TSPAN27

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

unconjugated

Quality Level

antibody product type

primary antibodies

clone

C33, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/ml

technique(s)

FUNC: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD82(3732)

General description

CD82 (KAI1), a member of the tetraspanin family, forms complexes with other tetraspanin proteins, integrins, coreceptors, MHC class I and II molecules. These complexes influence adhesion, morphology, activation, proliferation and differentiation of B, T and other cells. CD82 regulates cytoskeleton rearrangement and may participate in the turnover of the tetraspanin complex members. Besides in the plasma membrane, CD82 is localized also in endosome/lysosome compartments. Tumour-suppressive roles of CD82 have been demonstrated.

Specificity

The mouse monoclonal antibody C33 recognizes an extracellular/luminal epitope of CD82, a widely expressed cell surface protein of the tetraspanin family. CD82 is also found in endosome/lysosome compartments.

Immunogen

C91/PL (human HTLV-1+ T cell line)

Physical form

Phosphate buffered saline (PBS), pH 7.4

Preparation Note

Purified by protein-A affinity chromatography.

Storage and Stability

Store at 2-8°C. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service