Skip to Content
Merck
All Photos(2)

Documents

SAB4200481

Sigma-Aldrich

Anti-SATB1 antibody, Mouse monoclonal

clone SAT-5, purified from hybridoma cell culture

Synonym(s):

Anti-SATB homeobox 1, Anti-Special AT-rich sequence-binding protein 1, Monoclonal Anti-SATB1 antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

SAT-5, monoclonal

form

buffered aqueous solution

mol wt

antigen ~100 kDa

species reactivity

human, mouse, monkey, rat

concentration

~1.0 mg/mL

technique(s)

flow cytometry: 5-10 μg using THP-1 cells
immunocytochemistry: suitable
western blot: 2.0-4.0 μg/mL using Jurkat total cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SATB1(6304)

Related Categories

General description

Monoclonal Anti-SATB1 (mouse IgG2a isotype) is derived from the hybridoma SAT-5 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to a sequence at the N-terminus of human special AT-rich sequence-binding protein 1 (SATB1), conjugated to keyhole limpet hemocyanin (KLH). SATB1 encodes as nuclear matrix protein that interacts with scaffold-associating DNA. SATB1 gene is mapped to human chromosome 3p24.3.

Immunogen

a synthetic peptide corresponding to a sequence at the N-terminus of human SATB1, conjugated to KLH.

Application

Monoclonal Anti-SATB1 antibody produced in mouse has been used in:
  • immunoblotting
  • immunocytochemistry
  • flow cytometry

Biochem/physiol Actions

Special AT-rich sequence-binding protein 1 (SATB1) is a tumor promotor gene that plays an important role in regulating gene expression. The SATB1 known to be involved in organizing higher-order chromatin structure including the subnuclear organization of individual genes within multigene clusters in a tissue specific manner, locally and at a distance. It controls the global chromatin architecture via promyelocytic leukemia protein (PML). SATB1 also regulates the periodic anchoring of matrix attachment regions (MARs) in DNA to the nuclear matrix. This organizes chromatin into distinct loops. Furthermore, SATB1 recruits co-repressor complexes to its binding sites. SATB1 is abnormally expressed in human metastatic breast cancer and also promotes breast cancer tumor growth and metastasis. It is involved in the tumorigenesis of colorectal, coetaneous malignant melanoma and lung cancers. SATB1 also serves as a possible prognostic marker.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loss of special AT-rich binding protein 1 expression is a marker of poor survival in lung cancer
Selinger CJ, et al.
Journal of Thoracic Oncology : Official Publication of the International Association for the Study of Lung Cancer, 6(7), 1179-1189 (2011)
Acetylation-dependent interaction of SATB1 and CtBP1 mediates transcriptional repression by SATB1
Purbey PK, et al.
Molecular and Cellular Biology, 29(5), 1321-1337 (2009)
SATB1 downregulation induced by oxidative stress participates in trophoblast invasion by regulating $\beta$-catenin
Ra H, et al.
Biology of Reproduction, 98(6), 810-820 (2018)
SATB1 reprogrammes gene expression to promote breast tumour growth and metastasis
Han HJ, et al.
Nature, 452(7184), 187-187 (2008)
Correlation of SATB1 overexpression with the progression of human rectal cancer
Meng WJ, et al.
International Journal of Colorectal Disease, 27(2), 143-150 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service