Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200435

Sigma-Aldrich

Anti-COG7 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CDG2E, Anti-Component of oligomeric golgi complex 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~86 kDa

species reactivity

mouse, rat, human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysates of human HeLa cells.
indirect immunofluorescence: 1-2 μg/mL using human HeLa cells.
western blot: 2.5-5.0 μg/mL using whole extracts of mouse LA-4 and rat PC12 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COG7(91949)
mouse ... Cog7(233824)
rat ... Cog7(293456)

General description

Conserved Oligomeric Golgi complex 7 (COG7) protein belongs to the COG complex. COG complex is composed of eight distinct subunits organized in two heterotrimeric groups, COG2, 3, 4 and COG5, 6, 7 which are linked via a dimeric group formed by COG1 and COG8. COG complex is an evolutionarily conserved multi-subunit protein complex. The COG7 gene is located on the human chromosome at 16p12.2.

Specificity

Anti-COG7 recognizes human, mouse and rat COG7.

Immunogen

peptide corresponding to an internal region of human COG7, conjugated to KLH. The corresponding sequence is identical in mouse and rat COG7.

Application

Anti-COG7 antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Conserved Oligomeric Golgi complex (COG) modulates membrane trafficking and maintenance of Golgi glycosylation machinery in eukaryotic cells. COG7 protein is involved in retrograde vesicular trafficking. Mutations in the COG7 gene leads to type IIe congenital disorder of glycosylation.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Morava et al.
European journal of human genetics : EJHG, 15(6), 638-645 (2007-03-16)
We describe the clinical and biochemical characteristics in three patients from two different families diagnosed with Congenital Disorder of Glycosylation type IIe owing to a defect in Conserved Oligomeric Golgi complex (COG)7; one of the eight subunits of the COG.
Renate Zeevaert et al.
European journal of medical genetics, 52(5), 303-305 (2009-07-07)
We describe a patient homozygous for a novel mutation in COG7, coding for one of the subunits of the Conserved Oligomeric Golgi complex, involved in retrograde vesicular trafficking. His brother showed a similar clinical syndrome and glycosylation defect but no
G Charles Ostermeier et al.
Journal of andrology, 26(1), 70-74 (2004-12-22)
We recently described a complex population of spermatozoal coding RNAs that are delivered to the oocyte on fertilization. These are derived throughout spermatogenesis, representing a record of past events. Recently, evidence has been provided that micro-RNAs are present in testes
Rita Quental et al.
BMC evolutionary biology, 10, 212-212 (2010-07-17)
The Conserved Oligomeric Golgi (COG) complex is an eight-subunit assembly that localizes peripherally to Golgi membranes and is involved in retrograde vesicular trafficking. COG subunits are organized in two heterotrimeric groups, Cog2, -3, -4 and Cog5, -6, -7, linked by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service