Skip to Content
Merck
All Photos(3)

Key Documents

SAB2500308

Sigma-Aldrich

Anti-DGAT2 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DKFZp686A15125, Anti-Diacylglycerol O-acyltransferase homolog 2, Anti-GS1999full, Anti-HMFN1045

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
FACS
IHC
clone:
polyclonal
species reactivity:
canine, human, rat, mouse
citations:
4
technique(s):
ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

canine, human, rat, mouse

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DGAT2(84649)

General description

Diacylglycerol O-acyltransferase 2 (DGAT2) possesses one or two transmembrane domains and the amino- and carboxyl-terminal regions face the cytosol. The gene encoding it is localized on human chromosome 11q13.5 and has eight exons.

Immunogen

Peptide with sequence C-KLEHPTQQDIDLYH from the internal region (near the C Terminus) of the protein sequence according to NP_115953.2.

Biochem/physiol Actions

Diacylglycerol O-acyltransferase 2 (DGAT2) is involved in the final step of triacylglycerol biosynthesis. It esterifies diacylglycerol to triacylglycerol. Overexpression of the protein has been associated with hepatic steatosis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liver fat reduction with niacin is influenced by DGAT-2 polymorphisms in hypertriglyceridemic patients.
Hu M
Journal of Lipid Research (2012)
Roles of Acyl-CoA:Diacylglycerol Acyltransferases 1 and 2 in Triacylglycerol Synthesis and Secretion in Primary Hepatocytes.
Li C
Arteriosclerosis, Thrombosis, and Vascular Biology (2015)
Expressional profiling of prostate cancer risk SNPs at 11q13.5 identifies DGAT2 as a new target gene.
Nurminen R
Genes Chromosomes Cancer (2016)
Justin Chun et al.
Journal of the American Society of Nephrology : JASN, 33(5), 889-907 (2022-03-03)
Two variants in the gene encoding apolipoprotein L1 (APOL1) that are highly associated with African ancestry are major contributors to the large racial disparity in rates of human kidney disease. We previously demonstrated that recruitment of APOL1 risk variants G1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service