Skip to Content
Merck
All Photos(1)

Key Documents

SAB2101766

Sigma-Aldrich

Anti-PDSS2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BA59I9.3, Anti-C6orf210, Anti-DLP1, Anti-HDLP1, Anti-Prenyl (decaprenyl) diphosphate synthase, subunit 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

horse, guinea pig, rabbit, dog, rat, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDSS2(57107)

Immunogen

Synthetic peptide directed towards the C terminal region of human PDSS2

Biochem/physiol Actions

The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 (PDSS1; MIM 607429) and DLP1 (PDSS2) that produces Q10 ubiquinone.The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 (PDSS1; MIM 607429) and DLP1 (PDSS2) that produces Q10 ubiquinone (Saiki et al., 2005 [PubMed 16262699]).[supplied by OMIM]. Sequence Note: removed 1 base from the 5′ end that did not align to the reference genome assembly. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-24 DC341808.1 2-25 25-1292 BC039906.1 11-1278 1293-2534 AL832290.1 98-1339 2535-2535 BC039906.1 2521-2521 2536-3522 AL832290.1 1340-2326 3523-3568 AL832290.1 2328-2373

Sequence

Synthetic peptide located within the following region: IKAGKGVTSAIDLCRYHGNKALEALESFPPSEARSALENIVFAVTRFS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service