Skip to Content
Merck
All Photos(3)

Key Documents

SAB1405322

Sigma-Aldrich

Monoclonal Anti-LONRF1, (N-terminal) antibody produced in mouse

clone 1D9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ23749, RNF191

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D9, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.21 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LONRF1(91694)

General description

The gene LONRF1 (LON peptidase N-terminal domain and ring finger 1) is mapped to human chromosome 8p. It can be induced by androgens.

Immunogen

LONRF1 (NP_689484, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEESQSLNEPSPKQSEEIPEVTSEPVKGSLNRAQSAQSINSTEMPAREDCLKRVSSEPVLSVQEKGVLLKRKLSLLEQDVIVNEDGRNKLKKQGETPNEVCMFSLAYGDI

Biochem/physiol Actions

LONRF1 (LON peptidase N-terminal domain and ring finger 1) is a component of ubiquitin ligase complex and exhibits ATP-dependent peptidase activity. It might be associated with lymph node metastasis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su Tien Nguyen et al.
Cancer science, 98(5), 740-746 (2007-03-30)
An accurate assessment of the cervical lymph node metastasis status in oral cavity cancer not only helps predict the prognosis of patients, but also helps surgeons to perform the appropriate treatment. We investigated the utilization of microarray technology focusing on
Cheryl A Logan et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 300(6), R1373-R1383 (2011-03-18)
The capacities of eurythermal ectotherms to withstand wide ranges of temperature are based, in part, on abilities to modulate gene expression as body temperature changes, notably genes encoding proteins of the cellular stress response. Here, using a complementary DNA microarray
Vivek K Arora et al.
Cell, 155(6), 1309-1322 (2013-12-10)
The treatment of advanced prostate cancer has been transformed by novel antiandrogen therapies such as enzalutamide. Here, we identify induction of glucocorticoid receptor (GR) expression as a common feature of drug-resistant tumors in a credentialed preclinical model, a finding also
Hugo M Horlings et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 16(2), 651-663 (2010-01-14)
Several prognostic gene expression profiles have been identified in breast cancer. In spite of this progress in prognostic classification, the underlying mechanisms that drive these gene expression patterns remain unknown. Specific genomic alterations, such as copy number alterations, are an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service