Skip to Content
Merck
All Photos(1)

Documents

S9066

Sigma-Aldrich

SKF-89976A

>98% (HPLC), solid

Synonym(s):

1-(4,4-Diphenyl-3-butenyl)-3-piperidinecarboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25NO2 · HCl
CAS Number:
Molecular Weight:
371.90
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

>98% (HPLC)

form

solid

color

white

solubility

DMSO: soluble 20 mg/mL
H2O: insoluble

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

Cl[H].OC(=O)C1CCCN(CC\C=C(/c2ccccc2)c3ccccc3)C1

InChI

1S/C22H25NO2.ClH/c24-22(25)20-13-7-15-23(17-20)16-8-14-21(18-9-3-1-4-10-18)19-11-5-2-6-12-19;/h1-6,9-12,14,20H,7-8,13,15-17H2,(H,24,25);1H

InChI key

SNGGBKYQZVAQKA-UHFFFAOYSA-N

Application

SKF-89976A was used to study the role of adenosine receptors in uptake of GABA transport.4,5

Biochem/physiol Actions

SKF-89976A contains a cyclic amino acid with lipophilic moiety that enables the compound to cross the blood-brain barrier.1,2 It decreases the excitotoxic swelling of chick retina cells by blocking the glutamate-induced GABA release.3
GABA transporter type 1 (GAT-1) inhibitor that crosses the blood brain barrier.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anniina Alakuijala et al.
The European journal of neuroscience, 23(2), 514-520 (2006-01-20)
Both gamma-aminobutyric acid (GABA)(C) receptor subunit mRNA and protein are expressed in the stratum pyramidale in the CA1 area of the adult rat hippocampus, but so far no conclusive evidence about functional hippocampal GABA(C) receptors has been presented. Here, the
K E Andersen et al.
Journal of medicinal chemistry, 36(12), 1716-1725 (1993-06-11)
A series of different synthetic approaches to novel GABA uptake inhibitors are described, leading to examples which are derivatives of nipecotic acid and guvacine, substituted at nitrogen by 4,4-diaryl-3-butenyl or 2-(diphenylmethoxy)ethyl moieties. The in vitro value for inhibition of [3H]-GABA
G D Zeevalk et al.
Molecular and chemical neuropathology, 29(1), 27-36 (1996-09-01)
Acute excitotoxicity in the chick retina is characterized by cellular swelling and the subsequent selective release of GABA. In order to understand the source of GABA release, embryonic day 15 retina were incubated with 1 mM glutamate for 30 min
Andrea Soragna et al.
The Journal of physiology, 562(Pt 2), 333-345 (2004-10-30)
The relations between apparent affinity for substrates and operating rates have been investigated by two-electrode voltage clamp in the GABA transporter rGAT1 expressed in Xenopus oocytes. We have measured the transport current induced by the presence of GABA, as well
Annalisa Scimemi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 25(43), 10016-10024 (2005-10-28)
Persistent activation of GABAA receptors by extracellular GABA (tonic inhibition) plays a critical role in signal processing and network excitability in the brain. In hippocampal principal cells, tonic inhibition has been reported to be mediated by alpha5-subunit-containing GABAA receptors (alpha5GABAARs).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service