Skip to Content
Merck
All Photos(1)

Key Documents

S6381

Sigma-Aldrich

Sulfonazo III sodium salt

Synonym(s):

2,7-Bis(2-sulfophenylazo)chromotropic acid tetrasodium salt, 3,6-Bis(2-sulfophenylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic acid tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12N4Na4O14S4
CAS Number:
Molecular Weight:
776.57
Beilstein:
1900148
MDL number:
UNSPSC Code:
12171500

SMILES string

[Na+].[Na+].[Na+].[Na+].Oc1c(\N=N\c2ccccc2S([O-])(=O)=O)c(cc3cc(c(\N=N\c4ccccc4S([O-])(=O)=O)c(O)c13)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N4O14S4.4Na/c27-21-18-11(9-16(43(35,36)37)19(21)25-23-12-5-1-3-7-14(12)41(29,30)31)10-17(44(38,39)40)20(22(18)28)26-24-13-6-2-4-8-15(13)42(32,33)34;;;;/h1-10,27-28H,(H,29,30,31)(H,32,33,34)(H,35,36,37)(H,38,39,40);;;;/q;4*+1/p-4/b25-23+,26-24+;;;;

InChI key

FCMRBZBGGXLKBC-KBJWUIGUSA-J

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Sodium content on label.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service