Skip to Content
Merck
All Photos(1)

Key Documents

S0648

Sigma-Aldrich

Secretin porcine

~25 units/mg solid, from Porcine intestinal mucosa

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C130H220N44O41
CAS Number:
Molecular Weight:
3055.41
Beilstein:
741850
EC Number:
MDL number:
UNSPSC Code:
12352200

biological source

Porcine intestinal mucosa

specific activity

~25 units/mg solid

packaging

vial of ~100 Crick-Harper-Raper units

solubility

H2O: soluble
saline: soluble

UniProt accession no.

storage temp.

2-8°C

SMILES string

[H]N[C@@H](CC(O)=O)C(O)=O.[H]N[C@@H](CC(O)=O)C(O)=O.[H]N[C@@H](CC(O)=O)C(O)=O.[H]N[C@@H](CC(O)=O)C(O)=O.[H]N[C@@H](CC(O)=O)C(O)=O.[H]N[C@@H](CC(O)=O)C(O)=O.[H]N[C@@H](Cc1cnc[nH]1)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(O)=O)C(=O)NCC(=O)N[C@@H](C(C)O)C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C(C)C)C(N)=O

InChI

1S/C130H220N44O41.6C4H7NO4/c1-59(2)41-79(119(208)173-99(65(13)14)102(134)191)152-94(183)52-148-105(194)76(31-34-92(132)181)157-114(203)82(44-62(7)8)164-116(205)83(45-63(9)10)162-108(197)73(28-22-38-145-128(137)138)154-111(200)77(32-35-93(133)182)158-115(204)81(43-61(5)6)160-107(196)72(27-21-37-144-127(135)136)153-103(192)66(15)151-121(210)88(54-175)169-118(207)87(50-98(189)190)166-110(199)74(29-23-39-146-129(139)140)155-113(202)80(42-60(3)4)161-109(198)75(30-24-40-147-130(141)142)156-122(211)90(56-177)170-117(206)84(46-64(11)12)163-112(201)78(33-36-96(185)186)159-123(212)91(57-178)171-126(215)101(68(17)180)174-120(209)85(47-69-25-19-18-20-26-69)167-125(214)100(67(16)179)172-95(184)53-149-106(195)86(49-97(187)188)165-124(213)89(55-176)168-104(193)71(131)48-70-51-143-58-150-70;6*5-2(4(8)9)1-3(6)7/h18-20,25-26,51,58-68,71-91,99-101,175-180H,21-24,27-50,52-57,131H2,1-17H3,(H2,132,181)(H2,133,182)(H2,134,191)(H,143,150)(H,148,194)(H,149,195)(H,151,210)(H,152,183)(H,153,192)(H,154,200)(H,155,202)(H,156,211)(H,157,203)(H,158,204)(H,159,212)(H,160,196)(H,161,198)(H,162,197)(H,163,201)(H,164,205)(H,165,213)(H,166,199)(H,167,214)(H,168,193)(H,169,207)(H,170,206)(H,171,215)(H,172,184)(H,173,208)(H,174,209)(H,185,186)(H,187,188)(H,189,190)(H4,135,136,144)(H4,137,138,145)(H4,139,140,146)(H4,141,142,147);6*2H,1,5H2,(H,6,7)(H,8,9)/t66-,67+,68+,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,99-,100-,101-;6*2-/m0000000/s1

InChI key

KYSAMAXXLYMNME-INRVZKLKSA-N

Gene Information

pig ... SCT(397464)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

His-Ser-Asp-Gly-Thr-Phe-Thr-Ser-Glu-Leu-Ser-Arg-Leu-Arg-Asp-Ser-Ala-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Gly-Leu-Val-NH2

Biochem/physiol Actions

Stimulates secretion of carbonate-rich pancreatic fluid; relaxes smooth muscle

Preparation Note

Partially purified

Analysis Note

Activity determined by RIA using pure synthetic porcine secretin as standard.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuhisa Mori et al.
Pancreas, 42(2), 187-192 (2013-02-15)
The aim of this study was to establish a minimally invasive and simple screening test for detection of pancreatic ductal adenocarcinoma (PDAC) using duodenal juice (DJ). Duodenal juice was collected prospectively before endoscopic retrograde cholangiopancreatography in 46 patients. A protease
Maoqing Dong et al.
Regulatory peptides, 180, 5-11 (2012-11-13)
Understanding of the structural importance of each position along a peptide ligand can provide important insights into the molecular basis for its receptor binding and biological activity. This has typically been evaluated using serial replacement of each natural residue with
Revathi Sekar et al.
General and comparative endocrinology, 181, 18-24 (2012-12-19)
Secretin (Sct), traditionally a gastrointestinal hormone backed by a century long research, is now beginning to be recognized also as a neuroactive peptide. Substantiation by recent evidence on the functional role of Sct in various regions of the brain, especially
Xin-Yi Chen et al.
Neuroscience letters, 532, 29-32 (2012-11-15)
The paraventricular nucleus (PVN) of hypothalamus is a major integrative center in homeostatic control. Morphological studies have revealed a high level of secretin and secretin receptor expression in the PVN. To investigate the direct electrophysiological effects of secretin in the
Sophie S Abby et al.
PLoS genetics, 8(9), e1002983-e1002983 (2012-10-03)
Type 3 secretion systems (T3SSs) are essential components of two complex bacterial machineries: the flagellum, which drives cell motility, and the non-flagellar T3SS (NF-T3SS), which delivers effectors into eukaryotic cells. Yet the origin, specialization, and diversification of these machineries remained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service