Skip to Content
Merck
All Photos(1)

Key Documents

R2634

Sigma-Aldrich

BssH II from Bacillus stearothermophilus

buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

grade

for molecular biology

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Specificity

Recognition sequence: 5′-G/CGCGC-3′
Ligation and recutting results: After 2-10-fold BssH II overdigestion of 1 μg λ DNA substrate, results in 100% cutting, >95% of fragments can be ligated, and >95% recut.
Heat inactivation: Sensitive and inactivated at 80 °C for 20 minutes.
Comment: Optimal activity is at 50 °C

Other Notes

Supplied with 10x Restriction Endonuclease Buffer SA (B 7531).

Unit Definition

One unit is the enzyme activity that completely cleaves 1 μg λDNA in 1 hr. at 50 °C in a total volume of 25μl of buffer SA for restriction endonucleases.

Physical form

Solution in 10 mM Tris-HCl, pH 7.8, 50 mM KCl, 0.1 mM EDTA, 1 mM dithiothreitol, 500 μg/ml BSA, 50% glycerol (v/v) at 4 °C

incubation buffer

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Kessler et al.
Gene, 92(1-2), 1-248 (1990-08-16)
The properties and sources of all known class-I, class-II and class-III restriction endonucleases (ENases) and DNA modification methyltransferases (MTases) are listed and newly subclassified according to their sequence specificity. In addition, the enzymes are distinguished in a novel manner according
Nico Mitro et al.
Methods in molecular biology (Clifton, N.J.), 952, 137-144 (2012-10-27)
The role of certain amino acids in the interactions of ligands with their cognate nuclear receptors is usually achieved by the resolution of the crystal structure of the receptor complexed with the ligand. As a complementary functional approach, site-directed mutagenesis
Rachel M Smith et al.
Nucleic acids research, 41(1), 391-404 (2012-11-14)
Type IIB restriction-modification systems, such as BcgI, feature a single protein with both endonuclease and methyltransferase activities. Type IIB nucleases require two recognition sites and cut both strands on both sides of their unmodified sites. BcgI cuts all eight target
D S Jansson et al.
British poultry science, 53(6), 790-799 (2013-02-13)
1. Vancomycin-resistant Enterococcus faecium (VRE(fm)) has recently spread among Swedish broiler farms. The objectives were to investigate VRE(fm) persistence within barns between flocks, and to determine whether day-old chicks, feed or forklift trucks used for loading crates could be identified
Yu-Feng Huang et al.
BMC systems biology, 6 Suppl 2, S10-S10 (2013-01-11)
Current next-generation sequencing (NGS) platforms adopt two types of sequencing mechanisms: by synthesis or by ligation. The former is employed by 454 and Solexa systems, while the latter by SOLiD system. Although the pros and cons for each sequencing mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service