Skip to Content
Merck
All Photos(1)

Key Documents

R1156

Sigma-Aldrich

ROCK2 (5-554), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

KIAA0619, ROCK-II, ROKalpha

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

298-403 nmol/min·mg

mol wt

~88 kDa

UniProt accession no.

relevant disease(s)

cardiovascular diseases

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ROCK2(9475)

Biochem/physiol Actions

ROCK2 is a ubiquitously expressed serine/threonine kinase localized in the nucleus that regulates cytokinesis, smooth muscle contraction, the formation of actin stress fibers and focal adhesions, and the activation of the c-fos serum response element. ROCK2 is an immediate downstream target of the small GTPase RhoA. ROCK2 may play a pivotal role in cardiovascular diseases such as vasospastic angina, ischemic stroke, and heart failure. Inhibition of ROCKs by statins or other selective inhibitors leads to the upregulation and activation of endothelial nitric oxide synthase (eNOS) and reduction of vascular inflammation and atherosclerosis.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kensuke Noma et al.
American journal of physiology. Cell physiology, 290(3), C661-C668 (2006-02-14)
Rho-associated kinases (ROCKs), the immediate downstream targets of RhoA, are ubiquitously expressed serine-threonine protein kinases that are involved in diverse cellular functions, including smooth muscle contraction, actin cytoskeleton organization, cell adhesion and motility, and gene expression. Recent studies have shown
Hong Liu et al.
Molecular medicine reports, 19(5), 3882-3888 (2019-03-01)
MicroRNA‑122 (miR‑122) has been reported to be involved in the pathogenesis of several types of malignancies; however, its role in prostate carcinoma remains unknown. Thus, the current study aimed to investigate the functionality of miR‑122 in prostate carcinoma. Clinical data
Zhiyong Zhao et al.
Developmental biology, 275(1), 183-191 (2004-10-07)
Development of the endocardial cushions in the heart involves cell migration and cell differentiation, which is known as epithelial-mesenchymal transformation (EMT). These processes are regulated by cell signaling systems. Yet, the roles of intracellular GTPases and their effectors on these
Qing Zhou et al.
Hypertension (Dallas, Tex. : 1979), 69(6), 1070-1083 (2017-04-26)
Cardiac hypertrophy is characterized by increased myofibrillogenesis. Angiotensin II (Ang-II) is an essential mediator of the pressure overload-induced cardiac hypertrophy in part through RhoA/ROCK (small GTPase/Rho-associated coiled-coil containing protein kinase) pathway. FHOD3 (formin homology 2 domain containing 3), a cardiac-restricted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service