PZ0171
CP-100356 monohydrochloride
>98% (HPLC), powder, MDR1 (P-Gp) inhibitor
Synonym(s):
4-(3,4-Dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)-N-[2-(3,4-dimethoxyphenyl)ethyl]-6,7-dimethoxy-2-quinazolinamine monohydrochloride
About This Item
Recommended Products
product name
CP-100356 monohydrochloride, >98% (HPLC)
Assay
>98% (HPLC)
form
powder
storage condition
desiccated
color
white to off-white
solubility
DMSO: >5 mg/mL
storage temp.
2-8°C
SMILES string
Cl.COc1ccc(CCNc2nc(N3CCc4cc(OC)c(OC)cc4C3)c5cc(OC)c(OC)cc5n2)cc1OC
InChI
1S/C31H36N4O6.ClH/c1-36-24-8-7-19(13-25(24)37-2)9-11-32-31-33-23-17-29(41-6)28(40-5)16-22(23)30(34-31)35-12-10-20-14-26(38-3)27(39-4)15-21(20)18-35;/h7-8,13-17H,9-12,18H2,1-6H3,(H,32,33,34);1H
InChI key
WWCHXVYTCMPAMV-UHFFFAOYSA-N
Biochem/physiol Actions
Features and Benefits
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Discover Bioactive Small Molecules for ADME/Tox
Discover Bioactive Small Molecules for ADME/Tox
Discover Bioactive Small Molecules for ADME/Tox
Discover Bioactive Small Molecules for ADME/Tox
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service