Skip to Content
Merck
All Photos(1)

Key Documents

PCG3001

Sigma-Aldrich

TruPAGE TEA-Tricine SDS Running Buffer

20 ×

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NB.22

concentration

20 ×

compatibility

for use with Sigma-Aldrich TruPAGE Precast Gels

storage temp.

room temp

Application

TruPAGE TEA-Tricine SDS Running Buffer-20× has been used in Western blotting. [1][2]

Features and Benefits

TruPAGE TEA-Tricine SDS Running Buffer is specially formulated to be used with TruPAGE Precast Gels. This running buffer is ideally suited for resolution of a wide range of protein sizes. Please refer to the gel migration chart to select the appropriate gel and running buffer combination for your experimental needs.

Other Notes

TEA-Tricine SDS running buffer should be used if resolving proteins on 4–8% gels, which is ideal for larger molecular weight proteins.

Legal Information

TruPAGE is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Registered report: Transcriptional amplification in tumor cells with elevated c-Myc.
Blum D, et al.
eLife, 4 (2015)
Registered report: Oncometabolite 2-hydroxyglutarate is a competitive inhibitor of α-ketoglutarate-dependent dioxygenases.
Evans B, et al.
eLife, 4 (2015)
Eleanor Wendy Trotter et al.
Open biology, 10(10), 200200-200200 (2020-10-15)
Each approach used to synchronize cell cycle progression of human cell lines presents a unique set of challenges. Induction synchrony with agents that transiently block progression through key cell cycle stages are popular, but change stoichiometries of cell cycle regulators

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service