Assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
CC(O)C(NC(=O)C1CCCN1)C(=O)N2CCCC2C(=O)NC(CO)C(N)=O
Biochem/physiol Actions
Inhibits proteolysis of IgA by Neisseria gonorrheae protease type I
Other Notes
Analog of the hinge region of human IgA2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of medicinal chemistry, 32(10), 2407-2411 (1989-10-01)
The six series of unique tetrapeptides and their blocked N-acetyl, C-amide, and N-acetyl-C-amide analogues which comprise the hinge region of human IgA1 (Ser224 to Ser240) have been synthesized and tested as inhibitors of the type 1 IgA1 proteinase elaborated by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service