Skip to Content
Merck
All Photos(1)

Key Documents

P6237

Sigma-Aldrich

α-Pompilidotoxin

>98%

Synonym(s):

α-PMTX

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

>98%

form

solid

mol wt

1529

storage temp.

−20°C

Amino Acid Sequence

Arg-Ile-Lys-Ile-Gly-Leu-Phe-Asp-Gln-Leu-Ser-Lys-Leu

Biochem/physiol Actions

Voltage-gated sodium channel activator

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service