P2500
DL-Panthenol
Synonym(s):
(±)-α,γ-Dihydroxy-N-(3-hydroxypropyl)-β,β-dimethylbutyramide, (±)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide, DL-Pantothenyl alcohol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOCH2C(CH3)2CH(OH)CONH(CH2)3OH
CAS Number:
Molecular Weight:
205.25
Beilstein:
1724945
EC Number:
MDL number:
UNSPSC Code:
12352200
Recommended Products
mp
66-69 °C (lit.)
storage temp.
−20°C
SMILES string
CC(C)(CO)C(O)C(=O)NCCCO
InChI
1S/C9H19NO4/c1-9(2,6-12)7(13)8(14)10-4-3-5-11/h7,11-13H,3-6H2,1-2H3,(H,10,14)
InChI key
SNPLKNRPJHDVJA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service