P2186
[pGlu4]-Myelin basic protein fragment 4-14
≥97% (HPLC)
Synonym(s):
pGlu-Lys-Arg-Pro-Ser-Gln-Arg-Ser-Lys-Tyr-Leu
About This Item
Recommended Products
Assay
≥97% (HPLC)
form
powder
solubility
water: 5 mg/mL, clear, colorless
UniProt accession no.
storage temp.
−20°C
SMILES string
CC(C)CC(NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(CCCCN)NC(=O)C(CO)NC(=O)C(CCCNC(N)=N)NC(=O)C(CCC(N)=O)NC(=O)C(CO)NC(=O)C2CCCN2C(=O)C(CCCNC(N)=N)NC(=O)C(CCCCN)NC(=O)C3CCC(=O)N3)C(O)=O
InChI
1S/C60H100N20O17/c1-32(2)28-42(58(96)97)77-53(91)41(29-33-15-17-34(83)18-16-33)76-49(87)36(11-4-6-24-62)73-54(92)43(30-81)78-50(88)37(12-7-25-68-59(64)65)72-52(90)39(19-21-46(63)84)74-55(93)44(31-82)79-56(94)45-14-9-27-80(45)57(95)40(13-8-26-69-60(66)67)75-48(86)35(10-3-5-23-61)71-51(89)38-20-22-47(85)70-38/h15-18,32,35-45,81-83H,3-14,19-31,61-62H2,1-2H3,(H2,63,84)(H,70,85)(H,71,89)(H,72,90)(H,73,92)(H,74,93)(H,75,86)(H,76,87)(H,77,91)(H,78,88)(H,79,94)(H,96,97)(H4,64,65,68)(H4,66,67,69)
InChI key
RCRPGVVPXJBQFQ-UHFFFAOYSA-N
Gene Information
mouse ... Mbp(17196)
Amino Acid Sequence
Biochem/physiol Actions
Substrates
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service