Skip to Content
Merck
All Photos(1)

Key Documents

P1614

Sigma-Aldrich

Protein kinase Cζ pseudosubstrate, myristoyl trifluoroacetate salt

≥90% (HPLC)

Synonym(s):

PKC Zeta Pseudosubstrate Inhibitory peptide, ZIP

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€511.00

€511.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€511.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

€511.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥90% (HPLC)

form

powder

mol wt

calculated mol wt 1928

solubility

H2O: soluble

storage temp.

−20°C

Amino Acid Sequence

Myr-Ser-Ile-Tyr-Arg-Arg-Gly-Ala-Arg--Arg-Trp-Arg-Lys-Leu

Biochem/physiol Actions

Cell permeable protein kinase C (PKC) ζ inhibitor.

Features and Benefits

This compound is featured on the PKC page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P M Dang et al.
Journal of immunology (Baltimore, Md. : 1950), 166(2), 1206-1213 (2001-01-06)
Generation of superoxide anion by the multiprotein complex NADPH phagocyte oxidase is accompanied by extensive phosphorylation of its 47-kDa protein component, p47(phox), a major cytosolic component of this oxidase. Protein kinase C zeta (PKC zeta), an atypical PKC isoform expressed
Miguel Salinas-Saavedra et al.
eLife, 7 (2018-08-01)
In triploblastic animals, Par-proteins regulate cell-polarity and adherens junctions of both ectodermal and endodermal epithelia. But, in embryos of the diploblastic cnidarian Nematostella vectensis, Par-proteins are degraded in all cells in the bifunctional gastrodermal epithelium. Using immunohistochemistry, CRISPR/Cas9 mutagenesis, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service