Key Documents
OH30219114
Ohaus® pH Meter
Model ST300-G
About This Item
UNSPSC Code:
41115603
Product OH30219114 is not currently sold in your country. Contact Technical Service
material
epoxy shaft
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30219114
ORP measuring range
-1999-+1999 mV
pH measuring range
0.00-14.00 pH unit
temperature measuring range
0-100 °C
battery power
4 AAA, 250 hr life
General description
Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results. Ideal for secure field use with a meter stand, wrist strap and durable IP54 housing.
Application
pH, oxidation-reduction potential (ORP) measurement
Features and Benefits
Automatic and manual temperature compensation, auto buffer recognition, calibration icon
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)