Skip to Content
Merck
All Photos(1)

Key Documents

N8287

Sigma-Aldrich

NSC 625987

≥97% (HPLC)

Synonym(s):

1,4-Dimethoxy-9-thio(10H)-acridone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO2S
CAS Number:
Molecular Weight:
271.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

solubility

DMSO: 14 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(OC)c2C(=S)c3ccccc3Nc12

InChI

1S/C15H13NO2S/c1-17-11-7-8-12(18-2)14-13(11)15(19)9-5-3-4-6-10(9)16-14/h3-8H,1-2H3,(H,16,19)

InChI key

KFAKESMKRPNZTM-UHFFFAOYSA-N

Biochem/physiol Actions

NSC 625987 is a selective, potent Cdk4/cyclin D1 inhibitor.

Caution

Air-sensitive

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Kubo et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 5(12), 4279-4286 (2000-01-13)
Loss of p16 functional activity leading to disruption of the p16/cyclin-dependent kinase (CDK) 4:cyclin D/retinoblastoma pathway is the most common event in human tumorigenesis, suggesting that compounds with CDK4 kinase inhibitory activity may be useful to regulate cancer cell growth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service