Skip to Content
Merck
All Photos(1)

Key Documents

N8138

Sigma-Aldrich

Neuromedin U-25 porcine

≥97% (HPLC), solid

Synonym(s):

NMU-25

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C144H217N43O37
CAS Number:
Molecular Weight:
3142.53
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@@H]1CCCN1C(=O)CNC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)Cc3ccccc3)C(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc4ccc(O)cc4)C(=O)N[C@@H](Cc5ccccc5)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc6ccccc6)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N7CCC[C@H]7C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(N)=O)C(N)=O

InChI

1S/C144H217N43O37/c1-9-78(8)116(185-136(220)104-43-28-62-186(104)110(194)73-164-119(203)90(49-53-106(147)190)168-128(212)97(66-80-32-16-11-17-33-80)177-125(209)93(52-56-112(197)198)169-123(207)92(51-55-111(195)196)170-133(217)102(72-113(199)200)181-137(221)114(76(4)5)183-126(210)86(38-22-23-57-145)165-118(202)85(146)65-79-30-14-10-15-31-79)139(223)184-115(77(6)7)138(222)182-103(74-188)134(218)171-91(50-54-107(148)191)124(208)180-101(71-109(150)193)132(216)167-87(39-24-58-160-141(152)153)120(204)166-88(40-25-59-161-142(154)155)122(206)176-100(69-83-45-47-84(189)48-46-83)131(215)179-99(68-82-36-20-13-21-37-82)130(214)175-96(64-75(2)3)127(211)178-98(67-81-34-18-12-19-35-81)129(213)173-94(42-27-61-163-144(158)159)140(224)187-63-29-44-105(187)135(219)172-89(41-26-60-162-143(156)157)121(205)174-95(117(151)201)70-108(149)192/h10-21,30-37,45-48,75-78,85-105,114-116,188-189H,9,22-29,38-44,49-74,145-146H2,1-8H3,(H2,147,190)(H2,148,191)(H2,149,192)(H2,150,193)(H2,151,201)(H,164,203)(H,165,202)(H,166,204)(H,167,216)(H,168,212)(H,169,207)(H,170,217)(H,171,218)(H,172,219)(H,173,213)(H,174,205)(H,175,214)(H,176,206)(H,177,209)(H,178,211)(H,179,215)(H,180,208)(H,181,221)(H,182,222)(H,183,210)(H,184,223)(H,185,220)(H,195,196)(H,197,198)(H,199,200)(H4,152,153,160)(H4,154,155,161)(H4,156,157,162)(H4,158,159,163)/t78-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,114-,115-,116-/m0/s1

InChI key

YTYIPBDDJNZTTI-DXVWOPCVSA-N

Amino Acid Sequence

Phe-Lys-Val-Asp-Glu-Glu-Phe-Gin-Gly-Pro-Ile-Val-Ser-Gin-Asn-Arg-Arg-Tyr-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2

Biochem/physiol Actions

Potent stimulator of rat uterus smooth muscle.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Minamino et al.
Biochemical and biophysical research communications, 130(3), 1078-1085 (1985-08-15)
Two novel peptides eliciting a potent stimulant effect on the rat uterus smooth muscle have been purified and identified in porcine spinal cord. These peptides were designated as neuromedin U-8 (8 amino acids long) and U-25 (25 amino acids long)
T Kaisho et al.
International journal of obesity (2005), 41(12), 1790-1797 (2017-08-02)
Neuromedin U (NMU) is a neuropeptide with various physiological functions, including regulation of smooth-muscle contraction, blood pressure, stress responses and feeding behaviors. NMU activates two distinct receptors, NMUR1 and NMUR2, which are predominantly expressed in peripheral tissues and the central

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service