Skip to Content
Merck
All Photos(1)

Documents

N0254

Sigma-Aldrich

Ethyl nonadecanoate

~99%

Synonym(s):

Nonadecanoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H42O2
CAS Number:
Molecular Weight:
326.56
MDL number:
UNSPSC Code:
12352211

Assay

~99%

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCC(=O)OCC


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service