MTCH6800SK
MTC™ Bio Halo™ Lightweight Single Channel Pipettor Starter Kit
4 pipettors and a 6 place carousel stand
Synonym(s):
pipette
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio H6800-SK
Related Categories
General description
Halo Pipettor Starter Kit, includes H6800-10 (10µL), H6800-20 (20µL), H6800-200 (200µL), H680-1MXL (1250µL), and H6800-CR (6 place carousel stand)
Features and Benefits
- 10-40% lighter than leading competitors
- 4 Digit micrometer with DriftGuard protection
- Exclusive 1250µL model for extended length tips
- User-enabled calibration port
- Compatible with universal tips
Legal Information
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio