MTCA8000B
MTC™ Bio BowTie™ Floor standing Biohazard Bin With Lid
capacity × L 12-14 gal (45-52 L), W × L 25 in. (64 cm) × 35 in. (89 cm)
Synonym(s):
bio waste bag
Select a Size
€95.20
Select a Size
About This Item
€95.20
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio A8000B
W × L
25 in. (64 cm) × 35 in. (89 cm)
capacity × L
12-14 gal (45-52 L)
General description
Features and Benefits
- Drawstring for instant closure
- 12-16 gallon capacity, 25 x 35in.
- (45-60 liter capacity, 64 x 89cm)
- Color changing sterilization indicator
- Bags fit perfect in Biohazard Bin
- Bin has a hands-free foot pedal operated lid
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service