MSST0021
SILu™Prot CRP C-reactive protein human
recombinant, expressed in HEK 293 cells, 13C and 15N-labeled
Synonym(s):
PTX1, pentaxin, pentraxin
Select a Size
€679.00
Select a Size
About This Item
€679.00
Recommended Products
biological source
human
Quality Level
recombinant
expressed in HEK 293 cells
Assay
≥98% (SDS-PAGE)
form
liquid
potency
≥98% Heavy amino acids incorporation efficiency by MS
technique(s)
mass spectrometry (MS): suitable
suitability
suitable for mass spectrometry (standard)
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... PTX3(5806)
General description
Suggested Quantitative Analysis Parameters
(MRM settings provided for three suggested peptides)
Biochem/physiol Actions
Physical form
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service