Skip to Content
Merck
All Photos(1)

Key Documents

MAK534

Sigma-Aldrich

Salicylate Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€476.00

€476.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€476.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€476.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

application(s)

cosmetics

detection method

colorimetric

relevant disease(s)

rheumatological diseases

shipped in

ambient

storage temp.

2-8°C

General description

Salicylate is a salt or ester of salicylic acid, and can be found naturally in some plants. It is also a metabolic byproduct of aspirin (acetylsalicylic acid) and salicylate concentrations are often tested in blood or urine in cases of suspected overdose. Salicylic acid is commonly used in skincare products as an exfoliating ingredient, and in other consumer products as a preservative.

Features and Benefits

  • Linear detection range: 0.8 mM (10.9 mg/dL) to 20 mM (274.2 mg/dL)
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for the determination of salicylate in serum, plasma, urine, beauty products, mouthwash, etc.

Principle

The Salicylate Assay Kit provides a convenient and reliable means to measure salicylate. In the assay, salicylate complexes with ferric chloride to create a colored compound that can be measured at 560 nm.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service