Skip to Content
Merck
All Photos(1)

Key Documents

MAK529

Sigma-Aldrich

LDH Cytotoxicity Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

LDH-Cytotoxicity Test Kit II

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€161.00

€161.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€161.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€161.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

detection method

colorimetric

relevant disease(s)

hematological disorder; cancer

shipped in

wet ice

storage temp.

−20°C

General description

Lactate dehydrogenase (LDH) is an oxidoreductase which catalyzes the interconversion of lactate and pyruvate. Cytotoxic compounds often compromise cell membrane integrity by inducing apoptosis or necrosis. LDH is a stable cytosolic enzyme, that upon membrane damage, is released into the cellular environment. Therefore, LDH is often measured to evaluate the presence of tissue or cell damage.

Features and Benefits

  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for the rapid quantitative determination of cytotoxicity based on lactate dehydrogenase released into cell culture medium, and for the evaluation of toxic compounds, toxins, detergents, environmental pollutants and physical treatment on cell lysis.

Principle

The LDH Cytotoxicity Assay Kit is a simple and robust method to assess cytotoxic effects on cells by measuring the activity of LDH in cell culture supernatant. The assay is based on the reduction of a tetrazolium salt to a formazan dye.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service