Skip to Content
Merck
All Photos(2)

Key Documents

MAK212

Sigma-Aldrich

MMP-1 Inhibitor Screening Kit

Sufficient for 100 Fluorometric tests

Synonym(s):

Matrix Metalloproteinase-1 Inhibitor Screening Kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

detection method

fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

Gene Information

human ... MMP1(4312)

General description

Matrix Metalloproteinase-1 (MMP-1) is a secreted enzyme capable of breaking down interstitial collagen types I, II, and III in the extracellular matrix. Matrix metalloproteinases have important functions in morphogenesis, angiogenesis, apoptosis, tissue remodeling and repair, and tumor metastasis.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the screening of inhibitors of MMP-1.

Principle

The MMP-1 Inhibitor Screening Kit is a rapid, sensitive and high throughput assay to screen and characterize potential inhibitors of MMP-1. MMP-1 activity is measured by hydrolyzing a FRET-tagged substrate to yield a fluorescent product (λex = 490/λem = 520 nm) proportional to the enzymatic activity present.

Kit Components Only

Product No.
Description

  • MMP-1 Assay Buffer

  • MMP-1 Substrate

  • MMP-1 Enzyme

  • Inhibitor Control, GM6001

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service