Skip to Content
Merck
All Photos(1)

Key Documents

L8875

Sigma-Aldrich

L-Leucine ethyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)COOC2H5 · HCl
CAS Number:
Molecular Weight:
195.69
Beilstein:
3994312
EC Number:
MDL number:
UNSPSC Code:
12352200

storage temp.

2-8°C

SMILES string

Cl.CCOC(=O)[C@@H](N)CC(C)C

InChI

1S/C8H17NO2.ClH/c1-4-11-8(10)7(9)5-6(2)3;/h6-7H,4-5,9H2,1-3H3;1H/t7-;/m0./s1

InChI key

NOUDPBCEONUCOV-FJXQXJEOSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J Baker et al.
Macromolecular bioscience, 14(11), 1619-1626 (2014-08-27)
We report the encapsulation of three different proteases in bioinspired silica. Silica particles were formed under mild reaction conditions using cationic amine-rich ethyleneamines as initiators, which resulted in aggregations of nanoscale spheres. Following encapsulation, the proteases were characterized for their
David Schachter et al.
American journal of physiology. Gastrointestinal and liver physiology, 306(11), G938-G946 (2014-04-05)
The leucine-to-glutamate (Leu→Glu) pathway, which metabolizes the carbon atoms of l-leucine to form l-glutamate, was studied by incubation of rat tissue segments with l-[U-(14)C]leucine and estimation of the [(14)C]glutamate formed. Metabolism of the leucine carbon chain occurs in most rat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service