Skip to Content
Merck
All Photos(1)

Documents

L8257

Sigma-Aldrich

Lactoperoxidase from bovine milk

lyophilized powder (essentially salt-free), ≥80 units/mg protein

Synonym(s):

Donor:hydrogen peroxide oxidoreductase, Peroxidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder (essentially salt-free)

specific activity

≥80 units/mg protein

absorbance ratio

A412/280 nm 0.88-0.95

UniProt accession no.

storage temp.

−20°C

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

Gene Information

cow ... LPO(280844)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lactoperoxidase catalyzes the oxidation of iodide to iodine by hydrogen peroxide. This activity provides a gentle, specific alternative to chloramine T for the radioiodination of proteins and DNA.

Unit Definition

One unit will form 1.0 mg of purpurogallin from pyrogallol in 20 sec at pH 6.0 at 20 °C.

Analysis Note

Protein determined by Lowry method.

Other Notes

Using 2,2′-azinobis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt as substrate, two or three times the activity is observed.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lior Cohen et al.
The Journal of biological chemistry, 283(22), 15169-15176 (2008-03-15)
The bioactive surface of scorpion beta-toxins that interact with receptor site-4 at voltage-gated sodium channels is constituted of residues of the conserved betaalphabetabeta core and the C-tail. In an attempt to evaluate the extent by which residues of the toxin
A M Lew et al.
Proceedings of the National Academy of Sciences of the United States of America, 83(16), 6084-6088 (1986-08-01)
In addition to mRNA encoding the canonical form of the murine class I antigen H-2Kb (348 amino acid residues), mRNA that would encode a shortened form of H-2Kb (missing 9 amino acids from the C-terminus) has been identified in C57BL/6
Tracing with radiolabeled neurotrophins.
C S von Bartheld
Methods in molecular biology (Clifton, N.J.), 169, 195-216 (2001-01-06)
Karolina Haberska et al.
Talanta, 76(5), 1159-1164 (2008-09-02)
Lactoperoxidase (LPO) is an enzyme, which is used as an antimicrobial agent in a number of applications, e.g., food technology. In the majority of applications LPO is added to a homogeneous product phase or immobilised on product surface. In the
José Serrano et al.
Biochemical and biophysical research communications, 388(2), 443-449 (2009-08-15)
Oxidation of Amplex Red by hydrogen peroxide in the presence of horseradish peroxidase (HRP) gives rise to an intensely colour product, resorufin. This reaction has been frequently employed for measurements based on enzyme-coupled reactions that detect hydrogen peroxide as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service