Skip to Content
Merck
All Photos(1)

Key Documents

L7257

Sigma-Aldrich

Leucomalachite Green

Synonym(s):

Leucomalachite Green, p,p′-Benzylidenebis-N,N-dimethylaniline, 4,4′-Benzylidenebis(N,N-dimethylaniline)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[C6H4N(CH3)2]2
CAS Number:
Molecular Weight:
330.47
Beilstein:
2140506
EC Number:
MDL number:
UNSPSC Code:
12171500

mp

100-102 °C (lit.)

SMILES string

CN(C)c1ccc(cc1)C(c2ccccc2)c3ccc(cc3)N(C)C

InChI

1S/C23H26N2/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4/h5-17,23H,1-4H3

InChI key

WZKXBGJNNCGHIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service