Skip to Content
Merck
All Photos(2)

Documents

KEM0033

Sigma-Aldrich

T4 DNA Polymerase

Ultra-pure enzyme for nucleic acid modifications

Sign Into View Organizational & Contract Pricing


About This Item

grade

for molecular biology

Assay

>99% (SDS-PAGE)

form

buffered aqueous solution

specific activity

5,555 U/mg

concentration

3,000 U/mL

shipped in

dry ice

storage temp.

−20°C

General description

T4 DNA Polymerase catalyzes the extension of a primed DNA template in the 5′ → 3′ direction. This enzyme exhibits a powerful 3′ → 5′ exonuclease activity, while lacking any inherent 5′ → 3′ exonuclease or strand displacement functions.

Application

Suitable for DNA end-repair by fill-in of 5′ overhang and removal of 3′ overhang.

Features and Benefits

  • Ultra-purification process for ultimate enzyme performance
  • Highest quality specifications for ultimate product consistency
  • Undetectable DNA and nuclease contamination

Components

Supplied with:KEM0042B (10X Blue Buffer)

Unit Definition

One unit is defined as the amount of enzyme that will incorporate 10 nmol of dNTP into acid-precipitable material in 30 minutes at 37° C.

Physical form

Supplied in 100 mM KPO4, 1.0 mM DTT, 0.1 mM EDTA, and 50% glycerol at pH 6.5 @ 25° C.

Other Notes

Source of protein: Purified from a strain of E. coli that expresses the recombinant T4 DNA Polymerase gene.
Unit size: 2,000 U

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laure Rittié et al.
Journal of cell communication and signaling, 2(1-2), 25-45 (2008-09-04)
Since molecular cloning has become routine laboratory technique, manufacturers offer countless sources of enzymes to generate and manipulate nucleic acids. Thus, selecting the appropriate enzyme for a specific task may seem difficult to the novice. This review aims at providing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service