Skip to Content
Merck
All Photos(1)

Documents

K4393

Sigma-Aldrich

PKCι, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DXS11 79 E, PRKCl

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

564-764 nmol/min·mg

mol wt

~98 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKCI(5584)

General description

PKCι (protein kinase C ι) belongs to the protein kinase C family.

Biochem/physiol Actions

Protein kinase C ι (PKCι) is involved in Ras signaling. In colonic epithelium, it is an important regulator of oncogenic Ras. Transgenic mice which are constitutively expressing PKCι are at high risk of carcinogen-mediated colon carcinogenesis.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole R Murray et al.
The Journal of cell biology, 164(6), 797-802 (2004-03-17)
Protein kinase C iota (PKCiota) has been implicated in Ras signaling, however, a role for PKCiota in oncogenic Ras-mediated transformation has not been established. Here, we show that PKCiota is a critical downstream effector of oncogenic Ras in the colonic
J Rennecke et al.
European journal of biochemistry, 242(2), 428-432 (1996-12-01)
13 murine tissues and 12 cell lines were tested for the expression of the novel protein kinase C (PKC) isoenzyme mu. Using two different PKC mu antibodies (sc-639 and P26720), PKC mu was detected in all tissues and cells and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service