J4125
JKC 301
≥97% (HPLC)
Synonym(s):
Cyclo(D-Asp-Pro-D-Ile-Leu-D-Trp)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C32H44N6O7
CAS Number:
Molecular Weight:
624.73
MDL number:
UNSPSC Code:
12352200
Assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
CC[C@H](C)[C@H]1NC(=O)[C@@H]2CCCN2C(=O)[C@@H](CC(O)=O)NC(=O)[C@@H](Cc3c[nH]c4ccccc34)NC(=O)[C@H](CC(C)C)NC1=O
Biochem/physiol Actions
Endothelin receptor (ETA) antagonist
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service