I2875
DL-Indole-3-lactic acid
~98%
Synonym(s):
β-(3-Indolyl)lactic acid, 2-Hydroxy-3-(3-indolyl)propanoic acid, 3-(3-Indolyl)-2-hydroxypropanoic acid, 3-Indolyllactic acid, Indolelactic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H11NO3
CAS Number:
Molecular Weight:
205.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
~98%
mp
145-146 °C (lit.)
storage temp.
2-8°C
SMILES string
OC(Cc1c[nH]c2ccccc12)C(O)=O
InChI
1S/C11H11NO3/c13-10(11(14)15)5-7-6-12-9-4-2-1-3-8(7)9/h1-4,6,10,12-13H,5H2,(H,14,15)
InChI key
XGILAAMKEQUXLS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Reactant for preparation of:
- Antibacterial agents
- Dietary sweetener
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service