Skip to Content
Merck
All Photos(1)

Key Documents

I0881

Sigma-Aldrich

1,2-O-Isopropylidene-α-D-xylofuranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O5
CAS Number:
Molecular Weight:
190.19
Beilstein:
81520
MDL number:
UNSPSC Code:
12352200

storage temp.

−20°C

SMILES string

CC1(C)O[C@H]2O[C@H](CO)[C@H](O)[C@H]2O1

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arie Gruzman et al.
Journal of medicinal chemistry, 51(24), 8096-8108 (2008-12-04)
Type 2 diabetes mellitus has reached epidemic proportions; therefore, the search for novel antihyperglycemic drugs is intense. We have discovered that D-xylose increases the rate of glucose transport in a non-insulin-dependent manner in rat and human myotubes in vitro. Due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service