Key Documents
HS1846
Tungsten-point lab pen, for etching glass, ceramic and plastic
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
Product HS1846 is not currently sold in your country. Contact Technical Service
manufacturer/tradename
Heathrow Scientific HS1846
General description
Permanently mark glass slides, test tubes, and labware Sharp, tungsten carbide tip etches glass, ceramic, and plastic.
  • Retractable tip for safe storage
  • Measures 9.8 mm dia x 137 mm L
Choose from one of the most recent versions:
Certificates of Analysis (COA)